5-Acetyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

Supplier: Matrix Scientific

010491-500MG
101798-632EA 267.69 USD
101798-632
5-Acetyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
5-Acetyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₁NO₃
Melting Pt: 252…253 °C
MDL Number: MFCD00982313
CAS Number: 17106-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR