(5-Amino-4H-[1,2,4]triazol-3-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(5-Amino-1H-1,2,4-triazol-3-yl)acetic acid

010490-500MG
101798-630EA 246.15 USD
101798-630
(5-Amino-4H-[1,2,4]triazol-3-yl)acetic acid
(5-Amino-4H-[1,2,4]triazol-3-yl)acetic acid

Hazard: Irritant

Formula: C₄H₆N₄O₂
MW: 142.12 g/mol
Melting Pt: 220 °C (dec.)
MDL Number: MFCD00457974
CAS Number: 143832-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR