ɑ-Cyclohexyl-DL-mandelic acid
Supplier: Matrix Scientific
Synonyms:
α-Cyclohexylmandelic acid
, 2-Cyclohexyl-2-hydroxy-2-phenylacetic acid
010487-500MG
101798-624EA
267.69
USD
101798-624
ɑ-Cyclohexyl-DL-mandelic acid
alpha-Cyclohexyl-DL-mandelic acid
CAS: 4335-77-7
MDL Number: MFCD00194543
Molecular Formula: C14H18O3
Molecular Weight: 234,3
Hazard: Irritant
Formula:
C₁₄H₁₈O₃ MW: 234.30 g/mol Melting Pt: 161…163 °C |
MDL Number:
MFCD00194543 CAS Number: 4335-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...