3-Furan-2-yl-1-methylpropylamine

Supplier: Matrix Scientific

010458-500MG
101798-566EA 93.85 USD
101798-566
3-Furan-2-yl-1-methylpropylamine
3-Furan-2-yl-1-methylpropylamine

Hazard: Irritant

Formula: C₈H₁₃NO
Boiling Pt: 79 °C (90 Torr)
Density: 0.9629 (20 °C)
MDL Number: MFCD00520665
CAS Number: 768-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR