4-Benzylaniline hydrochloride

Supplier: Matrix Scientific

010434-500MG
101798-518EA 15.38 USD
101798-518
4-Benzylaniline hydrochloride
4-Benzylaniline hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₄ClN
Melting Pt: 219 °C
MDL Number: MFCD01679497
CAS Number: 6317-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR