1-(4-Fluorophenylsulfonyl)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
010431-500MG
101798-512EA
267.69
USD
101798-512
1-(4-Fluorophenylsulfonyl)-2-pyrrolidinecarboxylic acid
1-(4-Fluorophenylsulphonyl)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₂FNO₄S |
MDL Number:
MFCD00172808 CAS Number: 251096-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...