2,3-Dimethyl-1H-indole-5-carboxylic acid

Supplier: Matrix Scientific

010264-500MG
101798-190EA 461.54 USD
101798-190
2,3-Dimethyl-1H-indole-5-carboxylic acid
2,3-Dimethyl-1H-indole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
Melting Pt: 238…240 °C
MDL Number: MFCD00464053
CAS Number: 14844-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR