2,3-Dihydrobenzo[1,4]dioxine-6-sulfonyl chloride

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-6-sulphonyl chloride2,3-Dihydrobenzo[b][1,4]dioxine-6-sulfonyl chloride

010198-500MG
101798-068EA 61.54 USD
101798-068
2,3-Dihydrobenzo[1,4]dioxine-6-sulfonyl chloride
2,3-Dihydrobenzo[1,4]dioxine-6-sulfonyl chloride

Hazard: Irritant

Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
MDL Number: MFCD02677681
CAS Number: 63758-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR