2-Phenyl-3-(2-thienyl)acrylic acid

Supplier: Matrix Scientific

010196-500MG
101798-064EA 267.69 USD
101798-064
2-Phenyl-3-(2-thienyl)acrylic acid
2-Phenyl-3-(2-thienyl)acrylic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₂S
Melting Pt: 190…192 °C
MDL Number: MFCD01207139
CAS Number: 10569-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR