5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

010158-500MG
101797-990EA 267.69 USD
101797-990
5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid
5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂
MW: 222.6300 g/mol
Melting Pt: 250…251 °C
MDL Number: MFCD03715908
CAS Number: 54006-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR