3-Methylbenzofuran-2-carbonyl chloride

Supplier: Matrix Scientific

010146-500MG
101797-966EA 267.69 USD
101797-966
3-Methylbenzofuran-2-carbonyl chloride
3-Methylbenzofuran-2-carbonyl chloride

Hazard: Irritant

Formula: C₁₀H₇ClO₂
Boiling Pt: 1119…121 °C (4 Torr)
Melting Pt: 72…73 °C
MDL Number: MFCD01940394
CAS Number: 2256-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR