4-(Piperidin-1-yl)benzoic acid

Supplier: Matrix Scientific

010127-500MG
101797-926EA 24.62 USD
101797-926
4-(Piperidin-1-yl)benzoic acid
4-(Piperidin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MW: 205.2530 g/mol
Melting Pt: 222…224 °C
MDL Number: MFCD00587602
CAS Number: 22090-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR