2-Amino-5,5-dimethyl-5,6-dihydro-4H-benzothiazol-7-one

Supplier: Matrix Scientific

Synonyms: 2-Amino-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one

010115-500MG
101797-902EA 78.46 USD
101797-902
2-Amino-5,5-dimethyl-5,6-dihydro-4H-benzothiazol-7-one
2-Amino-5,5-dimethyl-5,6-dihydro-4H-benzothiazol-7-one

Hazard: Irritant

Formula: C₉H₁₂N₂OS
MW: 196.27 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00170187
CAS Number: 1629-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR