2-Amino-4-(1-naphthyl)thiazole

Supplier: Matrix Scientific

Synonyms: 4-(Naphthalen-1-yl)thiazol-2-amine

009921-500MG
101797-522EA 34.5 USD
101797-522
2-Amino-4-(1-naphthyl)thiazole
2-Amino-4-(1-naphthyl)thiazole

Hazard: Irritant

Formula: C₁₃H₁₀N₂S
MW: 226.30 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00236016
CAS Number: 56503-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR