1-Methyl-5-indoleboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methylindole-5-boronic acidN-Methylindole-5-boronic acid

009798-500MG
101797-290EA 15.38 USD
101797-290
1-Methyl-5-indoleboronic acid ≥95%
1-Methyl-5-indoleboronic acid

Hazard: Irritant/Refrigerate

Formula: C₉H₁₀BNO₂
MW: 174.9900 g/mol
MDL Number: MFCD03412070
CAS Number: 192182-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR