3-(Methylsulfonyl)phenylboronic acid ≥96%
Supplier: Matrix Scientific
Synonyms:
3-(Methylsulfonyl)benzeneboronic acid
, 3-(Methanesulphonyl)phenylboronic acid
, 3-(Methanesulfonyl)phenylboronic acid
, (3-(Methylsulphonyl)phenyl)boronic acid
, 3-(Methylsulphonyl)benzeneboronic acid
, (3-(Methylsulfonyl)phenyl)boronic acid
009795-1G
009795-5G
101797-280EA
60
USD
101797-280
101797-282
3-(Methylsulfonyl)phenylboronic acid ≥96%
3-(Methylsulphonyl)phenylboronic acid
Hazard: Irritant
Formula:
C₇H₉BO₄S MW: 200.02 g/mol Melting Pt: 102…110 °C |
MDL Number:
MFCD03092935 CAS Number: 373384-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...