4-Isoquinolineboronic acid ≥97%

Supplier: Matrix Scientific

009794-5G
101797-278EA 339.25 USD
101797-278
4-Isoquinolineboronic acid ≥97%
4-Isoquinolineboronic acid

Hazard: Irritant

Formula: C₉H₈BNO₂
MW: 172.9763 g/mol
MDL Number: MFCD03412093
CAS Number: 192182-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR