4-(Carboxymethyl)phenylboronic acid pinacol ester ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(Carboxymethyl)phenylboronic acid Pinacol ester
, 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid
, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
, 2-[(4-Carboxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
009783-500MG
101797-246EA
84.62
USD
101797-246
4-(Carboxymethyl)phenylboronic acid pinacol ester ≥97%
4-(Carboxymethyl)phenylboronic acid pinacol ester
Hazard: Irritant/Refrigerate
Formula:
C₁₄H₁₉BO₄ MW: 262.1093 g/mol |
MDL Number:
MFCD04038755 CAS Number: 797755-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...