Cyclohexylboronic acid pinacol ester ≥96%
Supplier: Matrix Scientific
Synonyms:
2-Cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohexane
, Cyclohexylboronic acid Pinacol ester
009774-1G
101797-232EA
109.23
USD
101797-232
Cyclohexylboronic acid pinacol ester ≥96%
Cyclohexylboronic acid pinacol ester
Hazard: Irritant
Formula:
C₁₂H₂₃BO₂ MW: 210.12 g/mol |
MDL Number:
MFCD04038749 CAS Number: 87100-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...