2-Quinolin-2-yl-1-[3-(trifluoromethyl)phenyl]ethanone

Supplier: Matrix Scientific

009301-1G
101796-822EA 89.23 USD
101796-822
2-Quinolin-2-yl-1-[3-(trifluoromethyl)phenyl]ethanone
2-Quinolin-2-yl-1-[3-(trifluoromethyl)phenyl]ethanone

Hazard: Irritant

Formula: C₁₈H₁₂F₃NO
MW: 315.3 g/mol
MDL Number: MFCD04037917
CAS Number: 849021-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR