2-(1,3-Benzoxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethanone

Supplier: Matrix Scientific

009300-1G 009300-5G
101796-818EA 92.31 USD
101796-818 101796-820
2-(1,3-Benzoxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethanone
2-(1,3-Benzoxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethanone

Hazard: Irritant

Formula: C₁₆H₁₀F₃NO₂
MW: 305.26 g/mol
MDL Number: MFCD04037916
CAS Number: 849021-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR