2-Quinolin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone

Supplier: Matrix Scientific

009294-1G
101796-802EA 100 USD
101796-802
2-Quinolin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone
2-Quinolin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone

Hazard: Irritant

Formula: C₁₈H₁₂F₃NO
MW: 315.3 g/mol
MDL Number: MFCD04037910
CAS Number: 283597-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR