1-Phenyl-2-quinoxalin-2-ylethanone

Supplier: Matrix Scientific

009288-1G
101796-790EA 129.23 USD
101796-790
1-Phenyl-2-quinoxalin-2-ylethanone
1-Phenyl-2-quinoxalin-2-ylethanone

Hazard: Irritant

Formula: C₁₆H₁₂N₂O
Melting Pt: 156…157 °C
MDL Number: MFCD00483316
CAS Number: 16310-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR