(R)-(-)-3-Fluoropyrrolidine hydrochloride ≥98%, ee 98%

Supplier: Matrix Scientific

Synonyms: (R)-(-)-3-Fluoropyrrolidine Hydrochloride

009061-1G
101796-240EA 78.46 USD
101796-240
(R)-(-)-3-Fluoropyrrolidine hydrochloride ≥98%, ee 98%
(R)-(-)-3-Fluoropyrrolidine hydrochloride

Hazard: Irritant

Formula: C₄H₉ClFN
MW: 125.57 g/mol
Melting Pt: 181…184 °C
MDL Number: MFCD04038717
CAS Number: 136725-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR