4-(Chlorodifluoromethyl)benzotrifluoride ≥97%
Supplier: Matrix Scientific
008869-1G
101795-670EA
173.85
USD
101795-670
4-(Chlorodifluoromethyl)benzotrifluoride ≥97%
4-(Chlorodifluoromethyl)benzotrifluoride
Hazard: Irritant
Formula:
C₈H₄ClF₅ |
CAS Number:
13947-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...