1-Bromo-3-(1,1,2,2-tetrafluoroethoxy)benzene ≥98%

Supplier: Matrix Scientific

008852-5G 008852-1G 008852-25G
101795-620EA 264.62 USD
101795-620 101795-616 101795-618
1-Bromo-3-(1,1,2,2-tetrafluoroethoxy)benzene ≥98%
1-Bromo-3-(1,1,2,2-tetrafluoroethoxy)benzene

Hazard: Irritant

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR