α-(Trifluoromethyl)benzhydrol ≥98%

Supplier: Matrix Scientific

008747-1G
101795-354EA 287.69 USD
101795-354
α-(Trifluoromethyl)benzhydrol ≥98%
α-(Trifluoromethyl)benzhydrol

Hazard: Irritant

Formula: C₁₄H₁₁F₃O
Boiling Pt: 157 °C (7 Torr)
Melting Pt: 74 °C
MDL Number: MFCD00021800
CAS Number: 379-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR