1-Benzyl-4,4-difluoropiperidine

Supplier: Matrix Scientific

008715-25G 008715-5G
101795-268EA 273.85 USD
101795-268 101795-266
1-Benzyl-4,4-difluoropiperidine
1-Benzyl-4,4-difluoropiperidine

Hazard: Irritant

Formula: C₁₂H₁₅F₂N
MDL Number: MFCD03840201
CAS Number: 155137-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR