4-Methoxy-3-(1H-pyrazol-1-ylmethyl)benzaldehyde

Supplier: Matrix Scientific

008612-500MG
101795-024EA 82.8 USD
101795-024
4-Methoxy-3-(1H-pyrazol-1-ylmethyl)benzaldehyde
4-Methoxy-3-(1H-pyrazol-1-ylmethyl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
CAS Number: 436086-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR