4-Piperidin-3-yl-5-(4-trifluoromethylphenyl)pyrimidin-2-ylamine

Supplier: Matrix Scientific

068224-250MG
101935-566EA 476.92 USD
101935-566
4-Piperidin-3-yl-5-(4-trifluoromethylphenyl)pyrimidin-2-ylamine
4-Piperidin-3-yl-5-(4-trifluoromethylphenyl)pyrimidin-2-ylamine

Hazard: Irritant

Formula: C₁₆H₁₇F₃N₄
MW: 322.34 g/mol
MDL Number: MFCD21606170
CAS Number: 1361112-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR