1-(1-Methanesulfonylpiperidin-4-ylmethyl)-1H-pyrazol-4-ylamine dihydrochloride

Supplier: Matrix Scientific

068216-500MG
101935-550EA 623.08 USD
101935-550
1-(1-Methanesulfonylpiperidin-4-ylmethyl)-1H-pyrazol-4-ylamine dihydrochloride
1-(1-Methanesulfonylpiperidin-4-ylmethyl)-1H-pyrazol-4-ylamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀Cl₂N₄O₂S
MDL Number: MFCD21606163
CAS Number: 1361116-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR