N-[2-(4-Aminopyrazol-1-yl)ethyl]-2-methoxyacetamide dihydrochloride

Supplier: Matrix Scientific

068208-500MG
101935-534EA 623.08 USD
101935-534
N-[2-(4-Aminopyrazol-1-yl)ethyl]-2-methoxyacetamide dihydrochloride
N-[2-(4-Aminopyrazol-1-yl)ethyl]-2-methoxyacetamide dihydrochloride

Hazard: Irritant

Formula: C₈H₁₆Cl₂N₄O₂
MW: 271.15 g/mol
MDL Number: MFCD21606157
CAS Number: 1361113-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR