7-Piperidin-2-yl-5-trifluoromethylimidazo[1,2-a]pyrimidine dihydrochloride

Supplier: Matrix Scientific

068206-250MG
101935-530EA 476.92 USD
101935-530
7-Piperidin-2-yl-5-trifluoromethylimidazo[1,2-a]pyrimidine dihydrochloride
7-Piperidin-2-yl-5-trifluoromethylimidazo[1,2-a]pyrimidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂F₃N₄
MW: 343.18 g/mol
MDL Number: MFCD21606155
CAS Number: 1361114-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR