6-(Morpholine-4-sulfonyl)indan-1-ylamine

Supplier: Matrix Scientific

068202-250MG
101935-522EA 476.92 USD
101935-522
6-(Morpholine-4-sulfonyl)indan-1-ylamine
6-(Morpholine-4-sulfonyl)indan-1-ylamine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃S
MW: 282.36 g/mol
MDL Number: MFCD21606151
CAS Number: 1337148-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR