(5-Piperidin-2-yl-2H-pyrazol-3-yl)methanol dihydrochloride

Supplier: Matrix Scientific

068190-250MG
101935-498EA 476.92 USD
101935-498
(5-Piperidin-2-yl-2H-pyrazol-3-yl)methanol dihydrochloride
(5-Piperidin-2-yl-2H-pyrazol-3-yl)methanol dihydrochloride

Hazard: Irritant

Formula: C₉H₁₇Cl₂N₃O
MDL Number: MFCD21606144
CAS Number: 1452547-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR