5-(3-Methylpiperidin-3-yl)-2H-pyrazol-3-ol dihydrochloride
Supplier: Matrix Scientific
068171-250MG
101935-460EA
687.69
USD
101935-460
5-(3-Methylpiperidin-3-yl)-2H-pyrazol-3-ol dihydrochloride
5-(3-Methylpiperidin-3-yl)-2H-pyrazol-3-ol dihydrochloride
Hazard: Irritant
Formula:
C₉H₁₇Cl₂N₃O MW: 254.16 g/mol |
MDL Number:
MFCD21606129 CAS Number: 1452566-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...