2-(6-Methylpyridin-2-yl)morpholine dihydrochloride

Supplier: Matrix Scientific

068163-250MG
101935-444EA 750.77 USD
101935-444
2-(6-Methylpyridin-2-yl)morpholine dihydrochloride
2-(6-Methylpyridin-2-yl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂O
MDL Number: MFCD21606121
CAS Number: 1361112-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR