2-Methyl-6-morpholin-2-ylpyridin-4-ol dihydrochloride

Supplier: Matrix Scientific

068161-250MG
101935-440EA 649.23 USD
101935-440
2-Methyl-6-morpholin-2-ylpyridin-4-ol dihydrochloride
2-Methyl-6-morpholin-2-ylpyridin-4-ol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂O₂
MDL Number: MFCD21606119
CAS Number: 1361115-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR