2-[2-(5-Bromopyrazin-2-yl)ethyl]piperidine-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

068157-250MG
101935-432EA 638.46 USD
101935-432
2-[2-(5-Bromopyrazin-2-yl)ethyl]piperidine-1-carboxylic acid tert-butyl ester
2-[2-(5-Bromopyrazin-2-yl)ethyl]piperidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₆H₂₄BrN₃O₂
MW: 370.3 g/mol
MDL Number: MFCD21606115
CAS Number: 1361114-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR