5-Methyl-4-piperidin-3-ylpyrimidin-2-ylamine dihydrochloride

Supplier: Matrix Scientific

068140-250MG
101935-398EA 638.46 USD
101935-398
5-Methyl-4-piperidin-3-ylpyrimidin-2-ylamine dihydrochloride
5-Methyl-4-piperidin-3-ylpyrimidin-2-ylamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈Cl₂N₄
MW: 265.19 g/mol
MDL Number: MFCD21606100
CAS Number: 1361114-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR