2-(5-Piperidin-3-yl-2H-pyrazol-3-yl)ethanol dihydrochloride

Supplier: Matrix Scientific

068136-250MG
101935-390EA 500 USD
101935-390
2-(5-Piperidin-3-yl-2H-pyrazol-3-yl)ethanol dihydrochloride
2-(5-Piperidin-3-yl-2H-pyrazol-3-yl)ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉Cl₂N₃O
MDL Number: MFCD21606096
CAS Number: 1452522-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR