3-(1',2',3',4',5',6'-Hexahydro[2,4']bipyridinyl-6-yl)-N,N-dimethylpropionamide dihydrochloride

Supplier: Matrix Scientific

068101-250MG
101935-320EA 687.69 USD
101935-320
3-(1',2',3',4',5',6'-Hexahydro[2,4']bipyridinyl-6-yl)-N,N-dimethylpropionamide dihydrochloride
3-(1',2',3',4',5',6'-Hexahydro[2,4']bipyridinyl-6-yl)-N,N-dimethylpropionamide dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₅Cl₂N₃O
MDL Number: MFCD21606070
CAS Number: 1361114-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR