4-(4-Methoxyphenyl)piperazine-1,2-dicarboxylic acid-1-tert-butyl ester

Supplier: Matrix Scientific

068093-250MG
101935-304EA 476.92 USD
101935-304
4-(4-Methoxyphenyl)piperazine-1,2-dicarboxylic acid-1-tert-butyl ester
4-(4-Methoxyphenyl)piperazine-1,2-dicarboxylic acid-1-tert-butyl ester

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₅
MDL Number: MFCD21606069
CAS Number: 1361111-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR