4-Pyridin-2-ylpiperazine-1,3-dicarboxylic acid-1-tert-butyl ester
Supplier: Matrix Scientific
068089-500MG
101935-296EA
615.38
USD
101935-296
4-Pyridin-2-ylpiperazine-1,3-dicarboxylic acid-1-tert-butyl ester
4-Pyridin-2-ylpiperazine-1,3-dicarboxylic acid-1-tert-butyl ester
Hazard: Irritant
Formula:
C₁₅H₂₁N₃O₄ MW: 307.35 g/mol |
MDL Number:
MFCD21606066 CAS Number: 1361111-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...