N-Methyl-3-(6-methyl-2-piperidin-3-ylpyrimidin-4-yl)propionamide dihydrochloride

Supplier: Matrix Scientific

068087-250MG
101935-292EA 638.46 USD
101935-292
N-Methyl-3-(6-methyl-2-piperidin-3-ylpyrimidin-4-yl)propionamide dihydrochloride
N-Methyl-3-(6-methyl-2-piperidin-3-ylpyrimidin-4-yl)propionamide dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₄Cl₂N₄O
MW: 335.28 g/mol
MDL Number: MFCD21606065
CAS Number: 1361113-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR