3-(2-Methyl-6-piperidin-4-ylpyrimidin-4-yl)propionamide

Supplier: Matrix Scientific

068073-250MG
101935-264EA 558.46 USD
101935-264
3-(2-Methyl-6-piperidin-4-ylpyrimidin-4-yl)propionamide
3-(2-Methyl-6-piperidin-4-ylpyrimidin-4-yl)propionamide

Hazard: Irritant

Formula: C₁₃H₂₀N₄O
MDL Number: MFCD18381619
CAS Number: 1316217-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR