2-(2,4-Dichlorobenzyl)-2H-indazole-3-carboxylic acid

Supplier: Matrix Scientific

068068-250MG
101935-254EA 476.92 USD
101935-254
2-(2,4-Dichlorobenzyl)-2H-indazole-3-carboxylic acid
2-(2,4-Dichlorobenzyl)-2H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀Cl₂N₂O₂
MDL Number: MFCD18381533
CAS Number: 123016-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR