[1,4]Oxazepane-6-carboxylic acid dimethylamide

Supplier: Matrix Scientific

068067-250MG
101935-252EA 647.69 USD
101935-252
[1,4]Oxazepane-6-carboxylic acid dimethylamide
[1,4]Oxazepane-6-carboxylic acid dimethylamide

Hazard: Irritant

Formula: C₈H₁₆N₂O₂
MDL Number: MFCD18381751
CAS Number: 1316223-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR