3-Chloro-6-[2-(1-methanesulfonylpyrrolidin-2-yl)ethyl]pyridazine

Supplier: Matrix Scientific

068063-250MG
101935-244EA 638.46 USD
101935-244
3-Chloro-6-[2-(1-methanesulfonylpyrrolidin-2-yl)ethyl]pyridazine
3-Chloro-6-[2-(1-methanesulfonylpyrrolidin-2-yl)ethyl]pyridazine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₂S
MDL Number: MFCD19691639
CAS Number: 1316225-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR