4-Chloro-2-(1-methanesulfonylpiperidin-3-yl)-6-methylpyrimidine

Supplier: Matrix Scientific

068061-250MG
101935-240EA 584.62 USD
101935-240
4-Chloro-2-(1-methanesulfonylpiperidin-3-yl)-6-methylpyrimidine
4-Chloro-2-(1-methanesulfonylpiperidin-3-yl)-6-methylpyrimidine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₂S
MDL Number: MFCD19691561
CAS Number: 1316218-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR